Learn More
Promocell GmbH PromoCell Pericyte Growth Medium 2 500 ml, ready-to-use with C-39841 attached to flask
Catalog No. NC1740096
Encompass
PromoCell Pericyte Growth Medium 2 has been developed for the in vitro cultivation of human pericytes from all tissues.In standard culture media pericytes tend to grow slowly and undergo early senescence, whereas in PromoCell Pericyte Growth Medium 2 the cells are optimally supported for cell proliferation and maintenance of multipotency.The Pericyte Growth Medium 2 is a low serum media and is more standardized version for an optimal culture.
Catalog No. NC1740096 Supplier Promocell GmbH Supplier No. C28041
PromoCell Pericyte Growth Medium 2 has been developed for the in vitro cultivation of human pericytes from all tissues.In standard culture media pericytes tend to grow slowly and undergo early senescence, whereas in PromoCell Pericyte Growth Medium 2 the cells are optimally supported for cell proliferation and maintenance of multipotency.The Pericyte Growth Medium 2 is a low serum media and is more standardized version for an optimal culture.