Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Peridox™ Concentrate Disinfectant
BIOPHARMA

Catalog No. 19033862 Shop All Contec Inc. Products
Change view
Click to view available options
Product Type:
Disinfectant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-033-862 Disinfectant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-033-862 Supplier Contec™ Supplier No. CR85342NK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peridox™ Concentrate is an EPA-registered disinfectant and cleaner that was designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens.

  • Broad spectrum disinfectant is fast acting and is effective against bacterial spores in 3 minutes, bacteria and viruses in 2 minutes, fungus in 1 minute and TB in 5 minutes.
  • Has more than 40 EPA registered claims
  • Does not contain bleach, alcohol, phenols or quaternary ammonium compounds.
  • Fast acting and will not damage stainless steel equipment
  • Compatible with all Contec mopping systems and hardware
  • Requires no rinsing and leaves no film on surfaces making it ideal for use on critical surfaces
  • With its shorter contact times and powerful wetting agents, Peridox Concentrate stays wet on the surface longer and works faster with shorter contact times and powerful wetting agents
  • Will not damage clothing, carpet, upholstery, LCD screens, and other surfaces when used as directed.
  • Patented, hyperactive chemistry provides fast results, 3-minute sporicide claim at 99.9999% efficacy
  • EPA-registered disinfectant and cleaner is ideal for use in life science cleanrooms
  • Concentrated formula provides cleanroom facilities with a cost-effective approach to use a high-level sporicidal disinfectant facility-wide
  • Compatible with most cleanroom materials of construction, and can be used on workstations, stainless steel equipment, floors, walls, and ceilings

Specifications

Product Type Disinfectant
pH 1.1 to 1.3
Sterility Non-sterile
For Use With (Application) Designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.