Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ PeridoxRTU™ Sporicidal Disinfectant
BIOPHARMA

Catalog No. 19033460 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (English):
32 oz.
1 gal.
Sterility:
Non-sterile
Sterile
Packaging:
Double bagged
Foaming trigger for Peridox disinfectant, Irradiated
Single bagged
Single bagged with push/pull cap (foaming trigger sold separately)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Packaging
19-033-460 32 oz. Non-sterile Single bagged with push/pull cap (foaming trigger sold separately)
19-033-461 1 gal. Non-sterile Single bagged
19-033-463 1 gal. Sterile Double bagged
19-168-543 32 oz. Non-sterile Foaming trigger for Peridox disinfectant, Irradiated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-033-460 Supplier Contec™ Supplier No. CR85365

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

PeridoxRTU ™ is a simple one-step cleaner, disinfectant and sporicide.

  • Broad spectrum disinfectant effective against bacterial spores, bacteria, viruses, TB and fungi (mold).
  • Excellent material compatibility that will not damage most surfaces in cleanroom environments, including stainless steel, acrylic, glass, aluminum and chrome.
  • Does not contain alcohol, bleach, quat or silver.
  • Daily cleaning and disinfecting tasks, such as wiping surfaces and mopping floors.
  • Effective component to monthly cleaning protocols.
  • PeridoxRTU is ideal for use in pharmaceutical, biopharmaceutical and medical device facilities.
  • Sterile PeridoxRTU is ideal for pharmaceutical and life science cleanroom environments, including sterile core areas.
  • Wear appropriate gloves and eye protection. Additional PPE may be required based on engineering controls/environment, method of application, and use protocols.

Order Info

Note: This part (19-033-460) is available in Fisher Canada only.

Specifications

pH 1.9 to 2.2 (at 20°C)
Sterility Non-sterile
Content And Storage Store in a cool, dry, well ventilated area away from sources of heat, combustible materials, direct sunlight, and incompatible substances. Keep container tightly closed when not in use. Maintain temperature between 0°C and 40°C. Do not allow product to freeze or become overheated. This may cause increased degradation of the product, which will decrease product effectiveness.
Dilution Ready to use. Do not dilute.
Disposable Consult with the US EPA Guidelines listed in 40 CFR Part 261.3 or the EU Directive 2008/98/EC on waste for the classifications of hazardous waste prior to disposal. Furthermore, consult with your state, local, or provincial waste requirements or guidelines, if applicable, to ensure compliance. Arrange disposal in accordance to the EPA and/or state and local guidelines.
Packaging Single bagged with push/pull cap (foaming trigger sold separately)
Volume (English) 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.