Learn More
Medchemexpress LLC HY-50909 100mg , Perifosine KRX-0401;NSC 639966;D21266 CAS:157716-52-4 Purity:98%
SDP
Catalog No. 501492939
Encompass_Preferred
Medchemexpress, HY-50909 100mg Perifosine KRX-0401;NSC 639966;D21266 CAS:157716-52-4 Formula:C25H52NO4P Akt Purity:98% Perifosine is an oral Akt inhibitor. All cells are sensitive to the antiproliferative properties of Perifosine with an IC 50 of ~0.6-8.9 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2939 Supplier Medchemexpress LLC Supplier No. HY50909100MG
Medchemexpress, HY-50909 100mg Perifosine KRX-0401;NSC 639966;D21266 CAS:157716-52-4 Formula:C25H52NO4P Akt Purity:98% Perifosine is an oral Akt inhibitor. All cells are sensitive to the antiproliferative properties of Perifosine with an IC 50 of ~0.6-8.9 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.