Learn More
Medchemexpress LLC HY-50909 50mg , Perifosine CAS:157716-52-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5090950MG
Encompass_Preferred
HY-50909 50mg Perifosine CAS: 157716-52-4 Perifosine is an oral Akt inhibitor which inhibits proliferation of different tumor cell lines with IC50s of 0.6-8.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0260