Learn More
Medchemexpress LLC Perifosine | 157716-52-4 | 99.9% | C25H52NO4P | 5 MG
SDP
Catalog No. 502029354
Encompass_Preferred
Perifosine is an oral Akt inhibitor that inhibits the proliferation of various tumor cell lines with IC50s ranging from 0.6 to 8.9 μM. It blocks cell cycle progression of head and neck squamous carcinoma cells at G1-S and G2-M phases by inducing p21WAF1, independent of p53 function. It also inhibits multiple signaling pathways in glial progenitors and can cooperate with temozolomide to arrest cell proliferation in gliomas in vivo. This product is for research use only and not sold to patients.
  • Inhibits proliferation of various tumor cell lines
  • Blocks cell cycle progression at G1-S and G2-M phases
  • Induces p21WAF1, independent of p53 function
  • Inhibits multiple signaling pathways in glial progenitors
  • Can cooperate with temozolomide to arrest cell proliferation in gliomas in vivo
Catalog No. 50-202-9354 Supplier Medchemexpress LLC Supplier No. HY509095MG
Perifosine is an oral Akt inhibitor that inhibits the proliferation of various tumor cell lines with IC50s ranging from 0.6 to 8.9 μM. It blocks cell cycle progression of head and neck squamous carcinoma cells at G1-S and G2-M phases by inducing p21WAF1, independent of p53 function. It also inhibits multiple signaling pathways in glial progenitors and can cooperate with temozolomide to arrest cell proliferation in gliomas in vivo. This product is for research use only and not sold to patients.
  • Inhibits proliferation of various tumor cell lines
  • Blocks cell cycle progression at G1-S and G2-M phases
  • Induces p21WAF1, independent of p53 function
  • Inhibits multiple signaling pathways in glial progenitors
  • Can cooperate with temozolomide to arrest cell proliferation in gliomas in vivo