Learn More
Selleck Chemical LLC Perifosine (KRX-0401) 5mg 157716-52-4 NSC639966
SDP
Catalog No. 508398
Encompass_Preferred
Perifosine (KRX-0401, NSC639966) is a novel Akt inhibitor with IC50 of 4.7 ?M in MM.1S cells, targets pleckstrin homology domain of Akt. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-839-8 Supplier Selleck Chemical LLC Supplier No. S10375MG
Perifosine (KRX-0401, NSC639966) is a novel Akt inhibitor with IC50 of 4.7 ?M in MM.1S cells, targets pleckstrin homology domain of Akt. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.