Learn More
OriGene Perilipin-1 (PLIN1) CytoSection 25 Slides
SDP
Catalog No. NC3734798
Encompass_Preferred
Transient overexpression of PLIN1, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3734798 Supplier OriGene Supplier No. TS406292P5
Transient overexpression of PLIN1, transcript variant 1, in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack