Learn More
Medchemexpress LLC Perilla ketone | 553-84-4 | MFCD01939833 | 99.9% | 166.22 | 10 MG
SDP
Catalog No. 5000425368
Encompass_Preferred
Perilla ketone is a naturally occurring xenobiotic compound. It is activated by pulmonary P450 cytochrome enzymes in the lung, which can lead to severe pulmonary damage and the development of diffuse pulmonary edema. It is for research use only.
  • Naturally occurring xenobiotic compound
  • Activated by pulmonary P450 cytochrome enzymes in the lung
  • Can lead to severe pulmonary damage and the development of diffuse pulmonary edema
  • For research use only
Catalog No. 50-004-25368 Supplier Medchemexpress LLC Supplier No. HYN950810MG
Perilla ketone is a naturally occurring xenobiotic compound. It is activated by pulmonary P450 cytochrome enzymes in the lung, which can lead to severe pulmonary damage and the development of diffuse pulmonary edema. It is for research use only.
  • Naturally occurring xenobiotic compound
  • Activated by pulmonary P450 cytochrome enzymes in the lung
  • Can lead to severe pulmonary damage and the development of diffuse pulmonary edema
  • For research use only