Learn More
Medchemexpress LLC (±)-Perillaldehyde | 2111-75-3 | MFCD00062990 | 96.7% | 150.22 g/mol | C10H14O | 5mg
SDP
Catalog No. 5000269308
Encompass_Preferred
( )-Perillaldehyde has an antidepressant effect by modulating the olfactory nervous system in a mouse model of stress-induced depression ( )-Perillaldehyde also has anti-inflammatory activity inducing JNK activation in RAW264 7 cells and inhibiting the expression of TNF- with an IC50 of 171 7 M[1][2]
Catalog No. 50-002-69308 Supplier Medchemexpress LLC Supplier No. HYW7537915MG
( )-Perillaldehyde has an antidepressant effect by modulating the olfactory nervous system in a mouse model of stress-induced depression ( )-Perillaldehyde also has anti-inflammatory activity inducing JNK activation in RAW264 7 cells and inhibiting the expression of TNF- with an IC50 of 171 7 M[1][2]