Learn More
Selleck Chemical LLC Perillaldehyde S3205-25mg
SDP
Catalog No. 503110503
Encompass_Preferred
Perillaldehyde (Perilladehyde Perillal PAE PA) the main component of Perilla frutescens (a traditional medicinal antioxidant herb) inhibits BaP-induced AHR activation and ROS production inhibits BaP/AHR-mediated release of the CCL2 chemokine and activats the NRF2/HO1 antioxidant pathway
Catalog No. 50-311-0503 Supplier Selleck Chemical LLC Supplier No. S320525MG
Perillaldehyde (Perilladehyde Perillal PAE PA) the main component of Perilla frutescens (a traditional medicinal antioxidant herb) inhibits BaP-induced AHR activation and ROS production inhibits BaP/AHR-mediated release of the CCL2 chemokine and activats the NRF2/HO1 antioxidant pathway