Learn More
Medchemexpress LLC HY-N2084 100mg Medchemexpress, Perillartine CAS:30950-27-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2084100MG
Encompass_Preferred
Medchemexpress, HY-N2084 100mg Perillartine CAS:30950-27-7 Perillartine is a sweetener, which activates the taste receptor type 1 member 2 (Tas1r2) subunit in a species-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3885