Learn More
Apexbio Technology LLC Perillartine 30950-27-7 200mg
SDP
Catalog No. 5000090072
Encompass_Preferred
Perillartine (CAS 30950-27-7) is a small molecule that modulates sweet taste perception by selectively interacting with the taste receptor type 1 member 2 (Tas1r2) subunit Studies indicate that Perillartine functions as a sweetener by activating Tas1r2 in a species-dependent manner thereby influencing taste signaling pathways This compound is primarily utilized in the development of food additives and in investigations of taste receptor function and chemosensory signaling mechanisms
Catalog No. 50-000-90072 Supplier Apexbio Technology LLC Supplier No. C7026200
Perillartine (CAS 30950-27-7) is a small molecule that modulates sweet taste perception by selectively interacting with the taste receptor type 1 member 2 (Tas1r2) subunit Studies indicate that Perillartine functions as a sweetener by activating Tas1r2 in a species-dependent manner thereby influencing taste signaling pathways This compound is primarily utilized in the development of food additives and in investigations of taste receptor function and chemosensory signaling mechanisms