Learn More
Cayman Chemical Perindopril, Purity: ≥95%, CAS: 82834-16-0, 100 mg
SDP
Catalog No. 502984279
Encompass_Preferred
An orally active ACE inhibitor that effectively lowers blood pressure; metabolized to perindoprilat, a specific, competitive inhibitor of ACE (IC50 = 1.5-3.2 nM in vitro)
Catalog No. 50-298-4279 Supplier Cayman Chemical Supplier No. 20796100 MG
An orally active ACE inhibitor that effectively lowers blood pressure; metabolized to perindoprilat, a specific, competitive inhibitor of ACE (IC50 = 1.5-3.2 nM in vitro)