Learn More
Electron Microscopy Sciences Periodic Acid, 0.5% Aqueous, 10450-60-9, 500ml
SDP
Catalog No. 5032035
Encompass_Preferred
Molecular Formula H5IO6, Molecular Weight 227.934, Periodic acid for the periodic acid Schiff, or PAS reaction, is used for the decetion of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. The material is treated with periodic acid Periodic acid creating aldehyde groups which react with Schiff's reagent and give a bright red color reaction, thus the nuclei appear blue.
Catalog No. 50-320-35 Supplier Electron Microscopy Sciences Supplier No. 1932405
Molecular Formula H5IO6, Molecular Weight 227.934, Periodic acid for the periodic acid Schiff, or PAS reaction, is used for the decetion of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. The material is treated with periodic acid Periodic acid creating aldehyde groups which react with Schiff's reagent and give a bright red color reaction, thus the nuclei appear blue.