Learn More
Newcomer Supply Periodic Acid 0.5%, Aqueous, 500 ml
SDP
Catalog No. NC0777902
Encompass
Use: PAS for glycogen or amoeba and in Gomori Basement Membrane Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC0777902 Supplier Newcomer Supply Supplier No. 13308B
Use: PAS for glycogen or amoeba and in Gomori Basement Membrane Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.