Learn More
Electron Microscopy Sciences Periodic Acid 0.5% Aqueous, 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2635603
Encompass_Preferred
Used in Puchtler-Sweat Method for Basement Membranes, Puchtler and Sweat (1964). References: Puchtler, H. and Sweat, F.: Stain Technology, 39:163, 1964 and AFIP Manual of Histological Staining Mehtods, 3rd ed., Ed. L. Luna: NewYork, McGraw Hill Pub., c. 1968, p. 73
Catalog No. 50-317-62