Learn More
Newcomer Supply Periodic Acid 1%, Aqueous, 500 ml
SDP
Catalog No. NC2101914
Encompass
Use: Can be diluted to 0.5% for PAS or PAMM. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101914 Supplier Newcomer Supply Supplier No. 13304A
Use: Can be diluted to 0.5% for PAS or PAMM. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.