Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Periodic Acid 1%
SDP

Catalog No. 21033451
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-451 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-451 Supplier StatLab Supplier No. PA16
Add to Cart
Add to Cart

Used to oxidize certain tissue elements to aldehydes prior to immersion in Schiff’s Reagent

  • Copilot Search Branding
  • Images
  • Videos
  • Periodic Acid-Schiff (PAS) staining process
  • Used primarily for the Periodic Acid-Schiff (PAS) staining process
  • This method is essential for detecting carbohydrates, glycogen, mucopolysaccharides, and glycoproteins in tissue samples
  • The staining process involves oxidizing carbohydrates to aldehydes, which then react with Schiff reagent to produce a magenta color, highlighting carbohydrate-rich structures in the tissue
  • This technique is invaluable for differentiating between normal and abnormal tissue components, offering essential clues for clinical diagnosis and research

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Periodic Acid
Quantity 16 oz.
Solution Type Oxidizer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.