Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic acid, ACS, 99.0-101.0%

Catalog No. AA3047218
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3047218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3047218 Supplier Thermo Scientific Chemicals Supplier No. 03047218
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in the oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White
Melting Point 126°C to 130°C
Sensitivity Hygroscopic; Light sensitive
Assay Percent Range 99.01-101.0%
Quantity 50 g
Physical Form Crystals
Solubility Information Soluble in water and alcohol.
Formula Weight 227.94
Grade ACS Reagent
Chemical Name or Material Periodic acid
Hazard Category H271-H314-H335-H372
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 233-937-0
RTECSNumber NN1269700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.