Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic Acid, MP Biomedicals™

Catalog No. MP210259590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210259590 500 g
MP210259525 25 g
MP210259580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP210259590 Supplier MP Biomedicals, Inc Supplier No. 0210259590
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for cell staining. Periodic Acid, MP Biomedicals stains the cytoplasm of cells found in the tubular pattern, mucus, and apical surfaces of various lumen and glycoproteins on SDS-PAGE gels. This compound can be used in oxidation reactions.

  • Periodic acid can be used in oxidation reactions converting sulfides into sulfones, catalyzed by Chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones
  • Periodic acid can also react with the 1,2 glycol linkage of carbohydrates forming aldehydes in tissue sections, which can be colored with Schiff bases, thus allowing for the staining of these tissues
  • Furthermore, studies show that periodic acid is not effective in staining cancer cells in solid nests, which are better stained using Grimelius
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

Melting Point 122°C (Lit.)
Boiling Point 135°C (literature)
Quantity 500 g
Physical Form Yellowish Crystalline Powder
Solubility Information Freely soluble in water (100mg/mL), alcohol, nitric acid; slightly soluble in ether.
Formula Weight 227.94
Percent Purity ≥99%
Recommended Storage Room Temperature (15–30°C). Store dessicated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.