Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic Acid, Reagent Grade, Reagents

Catalog No. 01337879
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-879 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-879 Supplier Reagents Holdings Llc Supplier No. C1267250100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Periodic Acid, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym Iodine Tetraoxide
IUPAC Name tetrahydroxyiodous acid
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White to Pale Yellow
Boiling Point 140°C
Packaging Glass Bottle
Specific Gravity 1.4
Quantity 100 g
Physical Form Solid
Grade Reagent
Chemical Name or Material Periodic Acid, Reagent Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.