Learn More
Ethos Biosciences Periodic Acid-Schiff Kit
Catalog No. 5000819198
Encompass_Preferred
Periodic Acid Schiff (PAS) reacts with aldehyde groups to produce a characteristic magenta color allowing visualization of glycogen mucopolysaccharides basement membranes and other carbohydrate-rich structures This kit includes Periodic Acid 0 5% Schiff s Reagent and Hematoxylin Provides consistent color development for reliable slide preparation and microscopic evaluation Suitable for routine and specialized PAS staining workflows supports accurate tissue and cellular analysis Supplied in chemically compatible containers it is appropriate for clinical research and educational laboratories
Catalog No. 50-008-19198 Supplier Ethos Biosciences Supplier No. 626216
Periodic Acid Schiff (PAS) reacts with aldehyde groups to produce a characteristic magenta color allowing visualization of glycogen mucopolysaccharides basement membranes and other carbohydrate-rich structures This kit includes Periodic Acid 0 5% Schiff s Reagent and Hematoxylin Provides consistent color development for reliable slide preparation and microscopic evaluation Suitable for routine and specialized PAS staining workflows supports accurate tissue and cellular analysis Supplied in chemically compatible containers it is appropriate for clinical research and educational laboratories