Learn More
Polysciences, Inc. Periodic Acid Schiff's (PAS) Stain Kit
SDP
Catalog No. NC9839645
Encompass
PAS techniques are used to demonstrate polysaccharides, neutral mucosubstances and basement membranes primarily in tissue. It is an easy, reproducible stain that provides exceptional stain quality. Kit also includes diastase powder to enable demonstration of glycogen by performing PAS with diastase digestion. Protocols available for both procedures.
Catalog No. NC9839645 Supplier Polysciences, Inc. Supplier No. 242001
PAS techniques are used to demonstrate polysaccharides, neutral mucosubstances and basement membranes primarily in tissue. It is an easy, reproducible stain that provides exceptional stain quality. Kit also includes diastase powder to enable demonstration of glycogen by performing PAS with diastase digestion. Protocols available for both procedures.