Learn More
Newcomer Supply Periodic Acid Schiff (PAS) Glycogen Control Slides, 98/set
SDP
Catalog No. NC1907203
Encompass
Newcomer Supply Periodic Acid Schiff (PAS) Glycogen Control Slides are for the positive histochemical staining of glycogen in tissue sections and can also be utilized as control slides for glycogen digestion steps. Periodic Acid Schiff (PAS) Glycogen Control Slides contain sections of positive staining liver.
Catalog No. NC1907203 Supplier Newcomer Supply Supplier No. 4540B
Newcomer Supply Periodic Acid Schiff (PAS) Glycogen Control Slides are for the positive histochemical staining of glycogen in tissue sections and can also be utilized as control slides for glycogen digestion steps. Periodic Acid Schiff (PAS) Glycogen Control Slides contain sections of positive staining liver.