Learn More
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit, 250 ml kit
SDP
Catalog No. NC2102284
Encompass
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit procedure, with methods for glycogen digestion, is used for staining glycoproteins and may aid in the differential diagnosis of tumors through detection of acid/neutral epithelial mucins and/or glycogen.   Digestion steps can be employed for further identification of mucosubstances.  PAS can also be used for staining basement membranes and fungal cell walls.
This item is not returnable. View return policy
Catalog No. NC2102284 Supplier Newcomer Supply Supplier No. 9162A
Newcomer Supply Periodic Acid Schiff (PAS) Stain Kit procedure, with methods for glycogen digestion, is used for staining glycoproteins and may aid in the differential diagnosis of tumors through detection of acid/neutral epithelial mucins and/or glycogen.   Digestion steps can be employed for further identification of mucosubstances.  PAS can also be used for staining basement membranes and fungal cell walls.