Learn More
Periodic Acid Solution, Sigma-Aldrich™
Catalog No. SD395132
Encompass
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
SD395132 1 L
1 options
Catalog No. SD395132 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3951321L
Reagent; Periodic Acid Solution; Sigma Aldrich; 1g/dL in deionized water (1% in deionized water); For use in Sigma Procedure No. 395 for Periodic Acid Schiff and Procedure No. HT100 for Silver Stain (Modified GMS); 1L
For in vitro diagnostic use
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 L