Learn More
Hand2Mind Periodic Table Connecting Tiles
SCI_ED
Catalog No. S00712
Change view
Click to view available options
Class Size:
1 Student
Catalog No. Class Size
S00712 1 Student
1 options
Catalog No. S00712 Supplier Hand2Mind Supplier No. 90634
Add to Cart
Add to Cart
Hands-on set that simplifies the learning of complex chemistry concepts in a fun and interactive way.
  • Hands-on set that simplifies the learning of complex chemistry concepts in a fun and interactive way
  • Unique plastic tiles snap together to form a complete periodic table of the elements.
  • Each 1 x 1 in. color-coded tile is printed with the atomic number, atomic symbol, atomic weight, and electron ring configuration for each of the elements
  • 163 tiles in 10 different colors
Specifications
Class Size 1 Student
Product Type Periodic Table Connecting Tiles