Learn More
Med Vet International Periosteotome Elevator, Stainless Steel, 2mm/4mm, 6in, J0899, Each
Catalog No. 5000585207
Encompass_Preferred
The periosteotome is a dual-ended instrument with flat-rounded tips and is used to gently elevate the gingival and mucosa away from a diseased pocket after an incision has been made. Tip widths are 2mm and 3mm. 6"L.
Catalog No. 50-005-85207 Supplier Med Vet International Supplier No. JOR899
The periosteotome is a dual-ended instrument with flat-rounded tips and is used to gently elevate the gingival and mucosa away from a diseased pocket after an incision has been made. Tip widths are 2mm and 3mm. 6"L.