Learn More
AdipoGen Periostin (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053336
Encompass_Preferred
Matched Pair Detection Set. Detects human periostin. Colorimetric assay. Sample Type: Cell Culture Supernatant, Plasma, Serum. Range: 80 to 5000pg/ml. Sensitivity: 30pg/ml. Periostin is also involved in many fundamental biological processes such as cell proliferation, cell invasion and angiogenesis. Changes in periostin expression are commonly detected in various cancers and pre-cancerous conditions, and periostin may be involved in regulating cancer cell activities that contribute to tumorigenesis, cancer progression and metastasis. Periostin up-regulation in cancers usually correlates with aggressiveness and/or poor survival. Periostin has shown to be involved in many aspects of allergic inflammation, such as eosinophil recruitment, airway remodeling, development of a Th2 phenotype and increased expression of inflammatory mediators. It is evaluated as a biomarker for bronchial asthma and airway inflammation.
Catalog No. 50-205-3336 Supplier AdipoGen Supplier No. AG46B0005KI01
Matched Pair Detection Set. Detects human periostin. Colorimetric assay. Sample Type: Cell Culture Supernatant, Plasma, Serum. Range: 80 to 5000pg/ml. Sensitivity: 30pg/ml. Periostin is also involved in many fundamental biological processes such as cell proliferation, cell invasion and angiogenesis. Changes in periostin expression are commonly detected in various cancers and pre-cancerous conditions, and periostin may be involved in regulating cancer cell activities that contribute to tumorigenesis, cancer progression and metastasis. Periostin up-regulation in cancers usually correlates with aggressiveness and/or poor survival. Periostin has shown to be involved in many aspects of allergic inflammation, such as eosinophil recruitment, airway remodeling, development of a Th2 phenotype and increased expression of inflammatory mediators. It is evaluated as a biomarker for bronchial asthma and airway inflammation.