Learn More
AdipoGen Periostin (human) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45B0004KI01
Encompass_Preferred
Periostin (human) ELISA Kit (96 wells), Adipogen Life Sciences. ELISA Assay. Detects natural and recombinant human periostin. Has been tested on human periostin isoforms 1 and 2 (should also detect human isoforms 3 and 4). Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum. Range: 78 to 5000pg/ml. Sensitivity: 15pg/ml. Periostin is also involved in many fundamental biological processes such as cell proliferation, cell invasion and angiogenesis. Periostin expression is increased by both transforming growth factor beta1 (TGF-beta1) and bone morphogenetic protein 2 (BMP-2). Periostin has shown to be involved in many aspects of allergic inflammation, such as eosinophil recruitment, airway remodeling, development of a Th2 phenotype and increased expression of inflammatory mediators. It is evaluated as a biomarker for bronchial asthma and airway inflammation.
Catalog No. 50-168-7262