Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Periostin ELISA Kit

Catalog No. EHPOSTNX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHPOSTNX5 5 x 96 Tests
EHPOSTN 96 Tests
EHPOSTNX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHPOSTNX5 Supplier Invitrogen™ Supplier No. EHPOSTNX5
Add to Cart
Add to Cart

ELISA

The Human Periostin ELISA quantitates Hu Periostin in human serum,plasma,or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PN. Principle of the method The Human Periostin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Periostin (PN), also designated osteoblast-specific factor 2 (OSF-2), is a disulfide linked protein originally isolated as a osteoblast-specific factor. Periostin is a secreted protein that binds heparin and functions as a ligand for alpha(V)beta(3) and alpha(V)beta(5) integrins. In preosteoblasts, Periostin acts as a cell adhesion molecule and plays a role in osteoblast recruitment, spreading and attachment. Periostin is mainly detected in lower gastrointestinal tract, aorta, stomach, placenta, uterus and breast tissues but is up-regulated in epithelial ovarian tumors and overexpressed in breast cancer. Expression of Periostin is increased by bone morphogenetic protein (BMP2) and transforming growth factor beta 1(TGF beta 1). Periostin contains a typical signal sequence, followed by a cysteine-rich domain, a fourfold repeated domain, which shows homology with the insect protein fasciclin, and a C-terminal domain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15063
Assay Range 0.082-60 ng/mL
Assay Sensitivity 80 pg/mL
Calibration Range 80 pg/mL to 60 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human Periostin. Other species not determined.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1A630052E07Rik; AI747096; fasciclin I-like; Fasciclin-I like; LOW QUALITY PROTEIN: periostin; MGC119510; MGC119511; OSF 2; Osf2; OSF-2; osteoblast specific factor 2; osteoblast specific factor 2 (fasciclin I-like); osteoblast-specific factor 2; PDLPOSTN; peri; periodontal ligament-specific periostin; periostin; periostin variant 1; periostin variant 2; periostin variant 3; periostin variant 4; periostin variant 5; periostin variant 6; periostin variant 7; periostin variant 8; periostin variant 9; periostin, osteoblast specific factor; periostin-like factor; Plf; PN; POSTN; POSTN protein; RP11 412K4.1; RP11-412K4.1; OSF-2; OSF2; PDLPOSTN; PN
Gene ID (Entrez) 10631
Gene Symbol Postn
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases PN, Osteoblast-specific factor 2, OSF-2
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym RP11-412K4.1, OSF-2, OSF2, PDLPOSTN, PN, periostin
Target Periostin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.