Learn More
AdipoGen Periostin mouse ELISA Kit
SDP
Supplier:  AdipoGen AG45B0005KI01
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects natural and recombinant mouse periostin. Has been tested on mouse periostin isoform 5 should also detect mouse isoforms 1, 2 and 3 according to sequences. Mouse isoform 4 has not been tested. Colorimetric assay. Sample Types Cell Culture Supernatant, Plasma, Serum. Range 31 to 2000pg/ml. Sensitivity 10pg/ml. Periostin is a 90-kDa matricellular protein that consists of a typical signal sequence, followed by a cysteine-rich region, an EMI domain protein-protein interactions, four tandem fasciclin-like domains that are responsible for integrin binding, and a C-terminal region. Periostin was originally isolated as an osteoblast-specific factor that functions as a cell adhesion molecule for pre-osteoblasts and in osteoblast recruitment, attachment and spreading. Periostin is also involved in many fundamental biological processes such as cell proliferation, cell invasion and angiogenesis.
Catalog No. 50-256-2740