Learn More
Medchemexpress LLC Periostin Protein M 100ug
SDP
Catalog No. 5000179240
Encompass_Preferred
Periostin protein is a multifunctional molecule that plays a key role in cellular processes by inducing cell attachment and spreading emphasizing its importance in cell adhesion dynamics Furthermore Periostin contributes to the structural integrity of connective tissue by enhancing the binding of BMP1 to the fibronectin matrix and promoting subsequent proteolytic activation of lysyl oxidase LOX Periostin Protein Mouse (HEK293 His) is the recombinant mouse-derived Periostin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79240 Supplier Medchemexpress LLC Supplier No. HYP73373100UG
Periostin protein is a multifunctional molecule that plays a key role in cellular processes by inducing cell attachment and spreading emphasizing its importance in cell adhesion dynamics Furthermore Periostin contributes to the structural integrity of connective tissue by enhancing the binding of BMP1 to the fibronectin matrix and promoting subsequent proteolytic activation of lysyl oxidase LOX Periostin Protein Mouse (HEK293 His) is the recombinant mouse-derived Periostin protein expressed by HEK293 with C-His labeled tag