Learn More
Med Vet International PerioWise Periodental Probe, J1340, 2/Pack
Catalog No. 5000584029
Encompass_Preferred
This probe has intense purple markings to provide optimal contrast and visibility in millimeter increments from 1-12mm. - Easier to quickly and accurately determine the depth of the pocket - Gentle to tissue, slightly flexible - Rounded tip for safety and comfort - Autoclavable and inexpensive - 2pk
Catalog No. 50-005-84029 Supplier Med Vet International Supplier No. JOR1340
This probe has intense purple markings to provide optimal contrast and visibility in millimeter increments from 1-12mm. - Easier to quickly and accurately determine the depth of the pocket - Gentle to tissue, slightly flexible - Rounded tip for safety and comfort - Autoclavable and inexpensive - 2pk