Learn More
StemExpress Peripheral Blood CD14+ Monocytes, Frozen, 10 million/vial
SDP
Supplier:  StemExpress PB14010C
Encompass
Human peripheral blood CD14+ monocytes are isolated from the mononuclear cell fraction by means of negative selection. Isolated cells are characterized by flow cytometry to ensure a highly pure and viable cell population.
Catalog No. 50-212-6367