Learn More
StemExpress Peripheral Blood CD3+ Pan T Cells, Frozen, 20 million/vial
SDP
Catalog No. 502126361
Encompass
Human peripheral blood CD3+ pan T cells are isolated by means of negative selection. Cells expressing CD14, CD15, CD16, CD19, CD36, CD56, CD123, and CD235a are depleted from the mononuclear cell population using immunomagnetic particles leaving purified, untouched CD3+ pan T cells. Isolated cells are characterized by flow cytometry to ensure a highly pure and viable cell population.
Catalog No. 50-212-6361 Supplier StemExpress Supplier No. PB03020C
Human peripheral blood CD3+ pan T cells are isolated by means of negative selection. Cells expressing CD14, CD15, CD16, CD19, CD36, CD56, CD123, and CD235a are depleted from the mononuclear cell population using immunomagnetic particles leaving purified, untouched CD3+ pan T cells. Isolated cells are characterized by flow cytometry to ensure a highly pure and viable cell population.