Learn More
Medchemexpress LLC HY-N1381 5mg Medchemexpress, Periplocin CAS:13137-64-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13815MG
Encompass_Preferred
Medchemexpress, HY-N1381 5mg Periplocin CAS:13137-64-9 Periplocin is a cardiotonic steroid isolated from Periploca forrestii. Periplocin promotes tumor cell apoptosis and inhibits tumor growth. Periplocin has the potential to facilitate wound healing through the activation of Src/ERK and PI3K/Akt pathways mediated by Na/K-ATPase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0333