Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Periplocoside N | 39946-41-3 | 1 MG
SDP

Catalog No. 5000417829
Encompass_Preferred

Periplocoside N is a pregnane glycoside isolated from the root powder of Periploca sepium. It exhibits insecticidal activities, specifically against the red imported fire ant.

  • Purity of 97.53%
  • Molecular weight is 464.63
  • Chemical formula is C27H44O6
  • Appears as a solid
  • Color is white to off-white
  • Classified as a steroid
  • Derived from plants
  • Acts as an inhibitor

Catalog No. 50-004-17829 Supplier Medchemexpress LLC Supplier No. HYN42501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Periplocoside N is a pregnane glycoside isolated from the root powder of Periploca sepium. It exhibits insecticidal activities, specifically against the red imported fire ant.

  • Purity of 97.53%
  • Molecular weight is 464.63
  • Chemical formula is C27H44O6
  • Appears as a solid
  • Color is white to off-white
  • Classified as a steroid
  • Derived from plants
  • Acts as an inhibitor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.