Learn More
Cayman Chemical Periplogenin, Purity: ≥98%, CAS: 514-39-6, 10 mg
SDP
Catalog No. 502911342
Encompass_Preferred
A cardenolide and an active metabolite of periplocin with diverse biological activities; reduces the activity of porcine Na+/K+ ATPase at 0.3 or 0.6 µM; induces apoptosis in U937 and PC3 prostate cancer cells, as well as inhibits the proliferation of these cells (EC50s = 1.41 and 1.2 µM, respectively) at 0.1 or 0.2 µM; increases ROS production and induces necroptosis in HaCaT keratinocytes at 2 µg/ml; reduces ear thickness in a mouse model of TPA-induced psoriasis
Catalog No. 50-291-1342 Supplier Cayman Chemical Supplier No. 3504710 MG
A cardenolide and an active metabolite of periplocin with diverse biological activities; reduces the activity of porcine Na+/K+ ATPase at 0.3 or 0.6 µM; induces apoptosis in U937 and PC3 prostate cancer cells, as well as inhibits the proliferation of these cells (EC50s = 1.41 and 1.2 µM, respectively) at 0.1 or 0.2 µM; increases ROS production and induces necroptosis in HaCaT keratinocytes at 2 µg/ml; reduces ear thickness in a mouse model of TPA-induced psoriasis