Learn More
Analytical Sales & Services Peristaltic Adapter for Semi-Rigid to Soft-Walled Tubing
Catalog No. 5000805101
Encompass
Peristaltic Adapter for 1/8” OD Semi-Rigid Tubing to 1.5-2.4mm ID Soft-Walled Tubing\n\nQty/Pkg: Each
Catalog No. 50-008-05101 Supplier Analytical Sales & Services Supplier No. 59798
Peristaltic Adapter for 1/8” OD Semi-Rigid Tubing to 1.5-2.4mm ID Soft-Walled Tubing\n\nQty/Pkg: Each