Learn More
New Era Pump Systems Inc The 9000 Series peristaltic pumps Rollers:4; Tubing ID 1/16""
Catalog No. nc0598738
Encompass_Preferred
The 9000 Series peristaltic pumps are equipped to be used in many settings from the small home based businesses to the large factory floor. Applications range from continuous flow to measured dispenses. The pump can be integrated into existing production lines or simply used stand alone.¿ The 9000 Series is fully programmable and can be operated manually or with a PC through a RS-232 communication port. PC cable and software offered as accessories. Rollers:4; Tubing ID 1/16""
Catalog No. NC0598738 Supplier New Era Pump Systems Inc Supplier No. NE9000G
The 9000 Series peristaltic pumps are equipped to be used in many settings from the small home based businesses to the large factory floor. Applications range from continuous flow to measured dispenses. The pump can be integrated into existing production lines or simply used stand alone.¿ The 9000 Series is fully programmable and can be operated manually or with a PC through a RS-232 communication port. PC cable and software offered as accessories. Rollers:4; Tubing ID 1/16""