Learn More
Instech Low-flow peristaltic pump, 900:1 motor delivers flow rates from 8-2400l/min.
SDP
Catalog No. 5081166
Encompass
P720 Peristaltic Pumps
Instech P720 peristaltic pumps deliver the low flow rates of syringe pumps without the limits on delivered volume. They are ideal for tissue perfusion and other low-flow laboratory applications.
The 900:1 motor delivers flow rates from 8-2400l/min depending on tube set size installed. Order tube sets separately. Universal power adapter included.
Features
  • Precise. Precision-machined pump heads, Swiss-made motor and gearhead assemblies, linear speed control, and a range of tube sizes deliver flow rates from 0.8 µL/min to 18 ml/min (0.05 to 1100 ml/hour) with an accuracy better than ±5%.
  • Small. The pumps fit in the palm of your hand and weigh about one pound.
  • Versatile. Pre-assembled tube sets are available with your choice of three tubing materials, nine types of connectors, and one or two channels.
Catalog No. 50-811-66 Supplier Instech Supplier No. P720
P720 Peristaltic Pumps
Instech P720 peristaltic pumps deliver the low flow rates of syringe pumps without the limits on delivered volume. They are ideal for tissue perfusion and other low-flow laboratory applications.
The 900:1 motor delivers flow rates from 8-2400l/min depending on tube set size installed. Order tube sets separately. Universal power adapter included.
Features
  • Precise. Precision-machined pump heads, Swiss-made motor and gearhead assemblies, linear speed control, and a range of tube sizes deliver flow rates from 0.8 µL/min to 18 ml/min (0.05 to 1100 ml/hour) with an accuracy better than ±5%.
  • Small. The pumps fit in the palm of your hand and weigh about one pound.
  • Versatile. Pre-assembled tube sets are available with your choice of three tubing materials, nine types of connectors, and one or two channels.