Learn More
Agilent Technologies PERISTALTIC PUMP TUBING 12/PK
Catalog No. NC2452300
Encompass
Peristaltic Pump Tubing for ICP-MS. Easy-Fit Peristaltic pump tubing, long-life, standard for spray chamber drain, 1 pack of 12 pcs. Beige thermoplastic elastomer, 3-stop, yellow/blue tabs, 1.52 mm id. Standard for pumping spray chamber drain to waste
Catalog No. NC2452300 Supplier Agilent Technologies Supplier No. 50050022
Peristaltic Pump Tubing for ICP-MS. Easy-Fit Peristaltic pump tubing, long-life, standard for spray chamber drain, 1 pack of 12 pcs. Beige thermoplastic elastomer, 3-stop, yellow/blue tabs, 1.52 mm id. Standard for pumping spray chamber drain to waste