Learn More
Perkin Elmer US LLC Silicone Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 6
Catalog No. NC9470521
Encompass_Preferred
This Silicone Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 6.
Catalog No. NC9470521 Supplier Perkin Elmer US LLC Supplier No. 00473552
This Silicone Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 6.