Learn More
Perkin Elmer US LLC PerkinElmer Capillary Column, Elite 624, 30 m, 0.25 mm I.D., 1.40 μm
Catalog No. NC3974880
Encompass
The Elite-624 column, featuring a low to mid-polarity (6%-cyanopropylphenyl)-dimethylpolysiloxane phase, is engineered for analyzing volatile organic pollutants and recommended in U.S. EPA methods. It achieves greater than 90% resolution for the first six gases in EPA Methods 8260 and 524.2, and is particularly effective for EPA Method 524.2 by resolving 2-nitropropane from 1,1-dichloropropanone. It is equivalent to USP G43 phase.
  • For use at temperatures from -20°C to 240°C.
  • Low to mid-polarity (6%-cyanopropylphenyl)-dimethylpolysiloxane phase.
  • Ideal for analyzing volatile organic pollutants and recommended in U.S. EPA methods.
  • Provides greater than 90% resolution of first six gases in EPA Methods 8260 and 524.2.
  • Resolves 2-nitropropane from 1,1-dichloropropanone for EPA Method 524.2.
  • Equivalent to USP G43 phase.
Catalog No. NC3974880 Supplier Perkin Elmer US LLC Supplier No. N9316201
Add to Cart
Add to Cart
The Elite-624 column, featuring a low to mid-polarity (6%-cyanopropylphenyl)-dimethylpolysiloxane phase, is engineered for analyzing volatile organic pollutants and recommended in U.S. EPA methods. It achieves greater than 90% resolution for the first six gases in EPA Methods 8260 and 524.2, and is particularly effective for EPA Method 524.2 by resolving 2-nitropropane from 1,1-dichloropropanone. It is equivalent to USP G43 phase.
  • For use at temperatures from -20°C to 240°C.
  • Low to mid-polarity (6%-cyanopropylphenyl)-dimethylpolysiloxane phase.
  • Ideal for analyzing volatile organic pollutants and recommended in U.S. EPA methods.
  • Provides greater than 90% resolution of first six gases in EPA Methods 8260 and 524.2.
  • Resolves 2-nitropropane from 1,1-dichloropropanone for EPA Method 524.2.
  • Equivalent to USP G43 phase.