Learn More
Perkin Elmer US LLC Consumables Kit for NexION 5000 Configuration
Supplier:  Perkin Elmer US LLC N8160027
Encompass_Preferred
PerkinElmer introduces the NexION 5000 ICP-MS Standard solution-ready sample introduction technology. It is engineered for the widest range of ICP-MS analyses. This kit includes: Fixed 2.0 mm Injector UHP Quartz Torch, Qty. 2 (N8152428) SilQ Cyclonic Spray Chamber with Matrix Gas Port, Qty. 1 (N8152539) Threaded Spray Chamber Drain Line, Qty. 2 (N8152456) Platinum Skimmer Cone for NexION 5000, Qty. 2 (N8161041) Platinum Sampler Cone, Qty. 2 (W1033614) Hyper Skimmer Cone for NexION 5000, Qty. 1 (N8160120) Sampler Cone Gasket, Qty. 3 (W1040148) Hyper Skimmer O-Ring, Qty. 2 (09902123) Gray/Gray Pump Tubing, Qty. 2 (N8152415) Flared PVC 0.51 mm Orange/Yellow Pump Tubing, Qty. 4 (N8152405) Drain Tubing (feet), Qty. 6 (02506495) Sample Uptake Tubing, Qty. 12 (WE024375) Sample Introduction Line for PFA-ST Nebulizer, Qty. 1 (N8152455) Female Barb CTFE Fitting, Qty. 4 (N8152388) Male Barb CTFE Fitting, Qty. 4 (N8152453) Instrument Filter (Left), Qty. 2 (N815216 Instrument Filter (Right), Qty.
Catalog No. 50-201-6407
Add to Cart
Request a Quote