Learn More
Fluotics PerkinElmer Janus compatible P30, filter/sterile
Catalog No. 5000435281
Encompass_Preferred
PerkinElmer Janus compatible P30, filter/sterile, Working Volume: 30uL, Tip Color: Clear, Tip Box, MDT, Compare To: 6001620, 50 racks/case with 384 tips per rack
Catalog No. 50-004-35281 Supplier Fluotics Supplier No. PKE30.F
PerkinElmer Janus compatible P30, filter/sterile, Working Volume: 30uL, Tip Color: Clear, Tip Box, MDT, Compare To: 6001620, 50 racks/case with 384 tips per rack