Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
GREENER_CHOICE

Catalog No. 14222448 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
25 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
14-222-448 200 μL Sterile Clear
14-222-442 175 μL Sterile Clear
14-222-443 25 μL Sterile Black
14-222-445 200 μL Non-sterile Black
14-222-444 25 μL Sterile Clear
14-222-441 175 μL Sterile Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-222-448 Supplier Axygen™ Supplier No. RRP200CRS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volume ranges from 20 to 200μL

  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.