Learn More
Fluotics PerkinElmer Janus Varispan compatible P1000, filter/sterile
Catalog No. 5000435168
Encompass
PerkinElmer Janus Varispan compatible P1000, filter/sterile, Working Volume: 950uL, Tip Color: Black, Hanging Rack, Varispan, Compare To: 6001258, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35168 Supplier Fluotics Supplier No. PKEH1000CB.F
PerkinElmer Janus Varispan compatible P1000, filter/sterile, Working Volume: 950uL, Tip Color: Black, Hanging Rack, Varispan, Compare To: 6001258, 24 racks/case with 96 tips per rack